270229
3-Methoxyphenethylamine
97%
Synonym(s):
2-(3-Methoxyphenyl)ethylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3OC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
151.21
Beilstein:
775202
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.538 (lit.)
bp
118-119 °C/6 mmHg (lit.)
density
1.038 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
COc1cccc(CCN)c1
InChI
1S/C9H13NO/c1-11-9-4-2-3-8(7-9)5-6-10/h2-4,7H,5-6,10H2,1H3
InChI key
WJBMRZAHTUFBGE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The spectral binding affinity for 3-methoxyphenethylamine was studied to examine the determinants of P450 2D6 catalysis.
Application
3-Methoxyphenethylamine was used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. It was also used in the synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tetrahedron, 62, 9002-9002 (2006)
Tetrahedron Letters, 48, 835-835 (2007)
P T Wong et al.
Neuroreport, 5(1), 53-56 (1993-10-25)
The effects of death and various treatments that affect the status of nigrostriatal neurones on striatal release of dopamine (DA) and 5-hydroxytryptamine (5-HT) and their metabolites 3,4-dihydroxyphenylacetic acid (DOPAC), 3-methoxy-4-hydroxyphenylethylamine (3-MT), homovanillic acid (HVA) and 5-hydroxyindoleacetic acid (5-HIAA) were studied
G P Miller et al.
Biochemistry, 40(47), 14215-14223 (2001-11-21)
Cytochrome P450 (P450) 2D6 oxidizes a wide variety of drugs typically at a distance of 5-7 A from a basic nitrogen on the substrate. To investigate the determinants of P450 2D6 catalysis, we analyzed the binding and oxidation of phenethylamine
Anita H Lewin et al.
Bioorganic & medicinal chemistry, 16(15), 7415-7423 (2008-07-08)
A cell line in which RD-HGA16 cells were stably transfected with the hTAAR 1 receptor was created and utilized to carry out a systematic evaluation of a series of beta-phenethylamines. Fair agreement was observed with data obtained for aryl and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service