Skip to Content
Merck
All Photos(2)

Key Documents

269980

Sigma-Aldrich

3-Phenoxyphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4OH
CAS Number:
Molecular Weight:
186.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.601 (lit.)

bp

185 °C/12 mmHg (lit.)

mp

41-42 °C (lit.)

density

1.159 g/mL at 25 °C (lit.)

SMILES string

Oc1cccc(Oc2ccccc2)c1

InChI

1S/C12H10O2/c13-10-5-4-8-12(9-10)14-11-6-2-1-3-7-11/h1-9,13H

InChI key

HBUCPZGYBSEEHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The thermodynamic properties of 3-phenoxyphenol was studied using a combination of effusion and calorimetric techniques and also by high-level ab initio molecular orbital calculations. The standard molar enthalpy of formation of 3-phenoxyphenol was determined using Calvet microcalorimetry[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel A V Ribeiro da Silva et al.
The Journal of organic chemistry, 76(10), 3754-3764 (2011-04-14)
Thermodynamic properties of 3- and 4-phenoxyphenol have been determined by using a combination of calorimetric and effusion techniques as well as by high-level ab initio molecular orbital calculations. The standard (p° = 0.1 MPa) molar enthalpies of formation in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service