Skip to Content
Merck
All Photos(1)

Key Documents

268151

Sigma-Aldrich

Triethylamine hydrochloride

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N · HCl
CAS Number:
Molecular Weight:
137.65
Beilstein:
3906409
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

Assay

98%

mp

261 °C (dec.) (lit.)

SMILES string

Cl.CCN(CC)CC

InChI

1S/C6H15N.ClH/c1-4-7(5-2)6-3;/h4-6H2,1-3H3;1H

InChI key

ILWRPSCZWQJDMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumanta Kumar Padhi et al.
Inorganic chemistry, 51(15), 8091-8102 (2012-07-26)
Cyclometalated ruthenium complexes having C(^)N and N(^)C type coordinating ligands with NAD(+)/NADH function have been synthesized and characterized by spectroscopic methods. The variation of the coordinating position of σ-donating carbon atom leads to a drastic change in their properties. Both
Akiko Ido et al.
Chemosphere, 90(1), 57-64 (2012-09-04)
Polychlorinated biphenyls (PCBs) were mainly used as lubricants and coolants in electrical equipment. However, their chemical stabilities as well as hydrophobic properties caused persistent environmental pollution and damage to human health based on their bioaccumulative property. PCBs are currently targeted
Juan M Padró et al.
Analytica chimica acta, 725, 87-94 (2012-04-17)
A new automated and rapid potentiometric method for determining the effect of organic-solvent composition on pK(a) has been developed. It is based on the measurements of pH values of buffer solutions of variable solvent compositions using a combined glass electrode.
Jun He et al.
Analytical chemistry, 84(12), 5236-5242 (2012-05-23)
By combining a novel chiral amino-acid surfactant containing an acryloyl amide tail, a carbamate linker, and a leucine headgroup of different chain lengths with a conventional cross-linker and a polymerization technique, a new "one-pot" synthesis for the generation of amino-acid
Eugenio Hardy et al.
Analytical biochemistry, 430(2), 203-213 (2012-09-11)
Nanoaggregates composed of selected glycoforms from Escherichia coli 055:B5 lipopolysaccharide (LPS) were prepared by combining sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis, zinc-imidazole reverse staining, zinc chelation after cutting gel slices, elution with either 0.5% triethylamine (TEA) or 0.4% to 0.5%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service