Skip to Content
Merck
All Photos(1)

Key Documents

268062

Sigma-Aldrich

D-Pipecolinic acid

99%

Synonym(s):

(R)-(+)-2-Piperidinecarboxylic acid, (R)-Piperidine-2-carboxylic acid, D-Homoproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
Beilstein:
81094
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]25/D +27°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

277 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H]1CCCCN1

InChI

1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)/t5-/m1/s1

InChI key

HXEACLLIILLPRG-RXMQYKEDSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodney A Fernandes et al.
Organic & biomolecular chemistry, 10(38), 7789-7800 (2012-08-23)
A newly developed π-allylpalladium with a (-)-β-pinene framework and an isobutyl side chain catalyzed the enantioselective allylation of imines in good yields and enantioselectivities (20 examples, up to 98% ee). An efficient enantioselective synthesis of the (R)-α-propyl piperonylamine part of
Hana Návarová et al.
The Plant cell, 24(12), 5123-5141 (2012-12-12)
Metabolic signals orchestrate plant defenses against microbial pathogen invasion. Here, we report the identification of the non-protein amino acid pipecolic acid (Pip), a common Lys catabolite in plants and animals, as a critical regulator of inducible plant immunity. Following pathogen
Kalyanasundaram Subramanian et al.
PloS one, 13(6), e0198990-e0198990 (2018-06-16)
D-amino acid oxidase (DAAO) degrades D-amino acids to produce α-ketoacids, hydrogen peroxide and ammonia. DAAO has often been investigated and engineered for industrial and clinical applications. We combined information from literature with a detailed analysis of the structure to engineer
Hai-Chao Xu et al.
Organic letters, 12(22), 5174-5177 (2010-10-16)
Catalytic intramolecular hydroamination of dithioketene acetals was developed for the synthesis of cyclic amino acid derivatives. Triggered by the addition of a catalytical amount of n-BuLi, the reaction proceeds to give proline and pipecolic acid derivatives in excellent yields and
Wen-Hua Chiou et al.
Organic & biomolecular chemistry, 10(13), 2518-2520 (2012-02-22)
Enantiomeric syntheses of (-)-homopipecolic acid and (-)-pelletierine have been achieved by chiral resolution of tropanol followed by Baeyer-Villiger oxidation. The methodology provides a practical route for the synthesis of optically pure piperidines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service