Skip to Content
Merck
All Photos(1)

Documents

266779

Sigma-Aldrich

Hafnium

wire, diam. 1.0 mm, ≥99.9% trace metals basis (purity excludes ~2% zirconium)

Synonym(s):

Celtium, Hafnium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Quality Level

Assay

≥99.9% trace metals basis (purity excludes ~2% zirconium)

form

wire

resistivity

29.6 μΩ-cm, 0°C

diam.

1.0 mm

bp

4602 °C (lit.)

mp

2227 °C (lit.)

density

13.3 g/cm3 (lit.)

SMILES string

[Hf]

InChI

1S/Hf

InChI key

VBJZVLUMGGDVMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

2 g = 20 cm

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurence Maggiorella et al.
Future oncology (London, England), 8(9), 1167-1181 (2012-10-04)
There is considerable interest in approaches that could improve the therapeutic window of radiotherapy. In this study, hafnium oxide nanoparticles were designed that concentrate in tumor cells to achieve intracellular high-energy dose deposit. Conventional methods were used, implemented in different
Daqin Chen et al.
Chemical communications (Cambridge, England), 48(86), 10630-10632 (2012-09-27)
Novel Yb/Er(Tm):Na(3)MF(7) (M = Zr, Hf) nanocrystals with intrinsic single-band upconversion emission, in contrast to the routine lanthanide-doped fluoride nanocrystals which show typical multi-band upconversion emissions, are reported for the first time. Specifically, the red upconversion intensity of the Yb/Er:Na(3)ZrF(7)
Hyun-June Jung et al.
Advanced materials (Deerfield Beach, Fla.), 24(25), 3396-3400 (2012-05-26)
Bismuth nanocrystals for a nanoscale floating gate memory device are self-assembled in Bi(2) Mg(2/3) Nb(4/3) O(7) (BMN) dielectric films grown at room temperature by radio-frequency sputtering. The TEM cross-sectional image shows the "real" structure grown on a Si (001) substrate.
José Luis Olivares-Romero et al.
Journal of the American Chemical Society, 134(12), 5440-5443 (2012-03-17)
Asymmetric epoxidation of allylic and homoallylic amine derivatives catalyzed by Hf(IV)-bishydroxamic acid complexes is described. Under similar conditions, aldimine and ketimine produced oxaziridines. The sulfonyl group is demonstrated to be an effective directing group for these transformations.
Suresh Kumar Raman Pillai et al.
ACS applied materials & interfaces, 4(12), 7047-7054 (2012-12-01)
The large-scale application of semiconducting single-walled carbon nanotubes (s-SWCNTs) for printed electronics requires scalable, repeateable, as well as noncontaminating assembly techniques. Previously explored nanotube deposition methods include serial methods such as inkjet printing and parallel methods such as spin-coating with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service