- Hf(IV)-catalyzed enantioselective epoxidation of N-alkenyl sulfonamides and N-tosyl imines.
Hf(IV)-catalyzed enantioselective epoxidation of N-alkenyl sulfonamides and N-tosyl imines.
Journal of the American Chemical Society (2012-03-17)
José Luis Olivares-Romero, Zhi Li, Hisashi Yamamoto
PMID22420598
ABSTRACT
Asymmetric epoxidation of allylic and homoallylic amine derivatives catalyzed by Hf(IV)-bishydroxamic acid complexes is described. Under similar conditions, aldimine and ketimine produced oxaziridines. The sulfonyl group is demonstrated to be an effective directing group for these transformations.