Skip to Content
Merck
All Photos(1)

Key Documents

265306

Sigma-Aldrich

2,3-Difluoroaniline

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$83.30

$83.30


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$83.30

About This Item

Linear Formula:
F2C6H3NH2
CAS Number:
Molecular Weight:
129.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$83.30


Estimated to ship onMay 26, 2025


Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.514 (lit.)

density

1.274 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Nc1cccc(F)c1F

InChI

1S/C6H5F2N/c7-4-2-1-3-5(9)6(4)8/h1-3H,9H2

InChI key

YCCQGFYAVUTQFK-UHFFFAOYSA-N

Application

2,3-Difluoroaniline was used in the synthesis of ethyl 2-arylamino-4-trifluoromethylpyrimidine-5-carboxylate[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solution-phase parallel synthesis using ion-exchange resins.
Tetrahedron, 54(16), 4141-4150 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service