Skip to Content
Merck
All Photos(2)

Key Documents

264156

Sigma-Aldrich

1-Chloro-9,10-bis(phenylethynyl)anthracene

99%

Synonym(s):

1-Chloro-BPEA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H17Cl
CAS Number:
Molecular Weight:
412.91
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

99%

form

powder or crystals

mp

203-205 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Clc1cccc2c(C#Cc3ccccc3)c4ccccc4c(C#Cc5ccccc5)c12

InChI

1S/C30H17Cl/c31-29-17-9-16-27-26(20-18-22-10-3-1-4-11-22)24-14-7-8-15-25(24)28(30(27)29)21-19-23-12-5-2-6-13-23/h1-17H

InChI key

IMMCAKJISYGPDQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jinhua Gan et al.
BioMed research international, 2020, 4510628-4510628 (2020-05-19)
Diabetic retinopathy (DR) is one of the hallmark complications of diabetes and a leading cause of vision loss in adults. Retinal pericyte death seems to be a prominent feature in the onset of DR. Pyroptosis is an inflammatory form of
P J Nicholls et al.
eNeuro, 9(5) (2023-01-14)
The protease caspase-3 is a key mediator of apoptotic programmed cell death. But weak or transient caspase activity can contribute to neuronal differentiation, axonal pathfinding, and synaptic long-term depression. Despite the importance of sublethal, or nonapoptotic, caspase activity in neurodevelopment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service