Skip to Content
Merck
All Photos(1)

Documents

263575

Sigma-Aldrich

2-Mercapto-5-nitrobenzimidazole

97%

Synonym(s):

5-Nitro-2-benzimidazolethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2S
CAS Number:
Molecular Weight:
195.20
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151614
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

274 °C (dec.) (lit.)

solubility

1 M NaOH: soluble 50 mg/mL, opaque, dark red

SMILES string

[O-][N+](=O)c1ccc2NC(=S)Nc2c1

InChI

1S/C7H5N3O2S/c11-10(12)4-1-2-5-6(3-4)9-7(13)8-5/h1-3H,(H2,8,9,13)

InChI key

YPXQSGWOGQPLQO-UHFFFAOYSA-N

General description

2-Mercapto-5-nitrobenzimidazole (MNB) is a charge transfer molecule and its influence on SiO2 and Au electrode on pentacene organic thin film transistors has been investigated. The treatment of the Au electrodes of an organic bottom-contact thin film transistor with MNB, improves the current-voltage characteristics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of simultaneous treatment of SiO2 gate and Au electrode with octadecyltrichlorosilane and charge transfer molecules on characteristics of pentacene thin film transistors.
Pyo KS and Song CK.
Thin Solid Films, 485(1), 230-234 (2005)
Influence of 2-mercapto-5-nitrobenzimidazole treatment on the electronic characteristics of bottom-contact organic field-effect transistors.
Park DS, et al.
Organic Electronics, 9(6), 1010-1016 (2008)
D R Doerge
Biochemistry, 25(16), 4724-4728 (1986-08-12)
The irreversible inactivation of bovine lactoperoxidase by thiocarbamide goitrogens was measured, and the kinetics were consistent with a mechanism-based (suicide) mode. Sulfide ion inactivated, 2-mercaptobenzimidazole-inactivated, and 1-methyl-2-mercaptoimidazole-inactivated lactoperoxidases have different visible spectra, suggesting different products were formed. The results support
Yuqing Gu et al.
Nature communications, 11(1), 516-516 (2020-01-26)
Anticounterfeiting labels based on physical unclonable functions (PUFs), as one of the powerful tools against counterfeiting, are easy to generate but difficult to duplicate due to inherent randomness. Gap-enhanced Raman tags (GERTs) with embedded Raman reporters show strong intensity enhancement and
Weidong Zhao et al.
Nanoscale, 12(35), 18056-18066 (2020-07-03)
An effective SERS substrate for on-field detection needs to satisfy high sensitivity to analyte and signal reproducibility even in the special case of tilting or bending of substrates. Herein, we transferred monolayer AuNPs into a nanocavity to construct a Au

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service