Skip to Content
Merck
All Photos(2)

Key Documents

301507

Sigma-Aldrich

2-Mercapto-1-methylimidazole

≥99%

Synonym(s):

Methimazole, 1-Methyl-2-imidazolethiol, 2-Mercapto-1-methylimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$148.00
25 G
$195.00

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$148.00
25 G
$195.00

About This Item

Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
Beilstein:
108646
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99%

mp

144-147 °C (lit.)

SMILES string

CN1C=CNC1=S

InChI

1S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3H,1H3,(H,5,7)

InChI key

PMRYVIKBURPHAH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Sensitive detection of 2-mercapto-1-methylimidazole by Au-Ag-Au double shell nanoparticles-based localized surface plasmon resonance and surface-enhanced Raman scattering biosensor was reported.[1]

Application

2-Mercapto-1-methylimidazole was employed as hydrophobic charge-induction chromatography ligand for antibody purification.[2] It was also used in preparation of nitrile functionalized methimazole-based room temperature ionic liquids.[3]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Li Lu et al.
Journal of chromatography. A, 1278, 61-68 (2013-01-23)
Hydrophobic charge-induction chromatography (HCIC) is a novel technology for antibody purification. The ligand densities and pore properties of HCIC resins have significant effects on the separation behavior of protein, however, the understandings are quite limited. In the present work, new
Xue Liao et al.
Talanta, 117, 203-208 (2013-11-12)
In this paper, Au-Ag-Au double shell nanoparticles were prepared based on the reduction of the metal salts HAuCl4 and AgNO3 at the surface of seed particles. Due to the synergistic effect between Au and Ag, the hybrid nanoparticles are particularly
Amal I Siriwardana et al.
The Journal of organic chemistry, 75(24), 8376-8382 (2010-11-18)
The alkylation reaction of 2-mercapto-1-methylimidazole 1b with 2-chloroacetonitrile and 2-chloropropionitrile produced S-alkyl methimazole chlorides 2a and 2b which were subjected to anion metathesis with lithium bis(trifluoromethanesulfonyl)amide, LiNTf(2), to afford nitrile functionalized methimazole-based room temperature ionic liquids 3a and 3b in
Coralie Leblicq et al.
Pediatrics, 131(2), e616-e619 (2013-01-09)
A 15-year-old girl presented with chorea as a first sign of Graves' hyperthyroidism. Chorea abated with antithyroid drug treatment and reappeared when hyperthyroidism recurred but not when thyrotropin receptor antibodies increased after administration of (131)I. Therefore, chorea in this patient
F S Boretti et al.
Journal of veterinary internal medicine, 27(2), 377-381 (2013-02-13)
Transdermal methimazole is an acceptable alternative to oral treatment for hyperthyroid cats. There are, however, no studies evaluating the duration of T4 suppression after transdermal methimazole application. Such information would be valuable for therapeutic monitoring. To assess variation in serum

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service