Skip to Content
Merck
All Photos(1)

Key Documents

262420

Sigma-Aldrich

2,6-Dimethoxybenzoyl chloride

80%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCl
CAS Number:
Molecular Weight:
200.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

mp

64-66 °C (lit.)

SMILES string

COc1cccc(OC)c1C(Cl)=O

InChI

1S/C9H9ClO3/c1-12-6-4-3-5-7(13-2)8(6)9(10)11/h3-5H,1-2H3

InChI key

NDXRPDJVAUCBOH-UHFFFAOYSA-N

Application

2,6-Dimethoxybenzoyl chloride has been used in the preparation of:
  • 2,6-dimethoxyphenylpenicillin[1]
  • didechloro-OO-dimethylpyoluteorin[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

274. Derivatives of 6-aminopenicillanic acid. Part III. 2, 6-Dialkoxybenzoyl derivatives.
Doyle FP, et al.
Journal of the Chemical Society, 1453-1458 (1962)
506. The structure of pyoluteorin.
Birch AJ, et al.
Journal of the Chemical Society, 2641-2644 (1964)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service