Skip to Content
Merck
All Photos(1)

Key Documents

288365

Sigma-Aldrich

3,4-Difluoronitrobenzene

99%

Synonym(s):

1,2-Difluoro-4-nitrobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$145.00

$145.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$145.00

About This Item

Linear Formula:
F2C6H3NO2
CAS Number:
Molecular Weight:
159.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$145.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.509 (lit.)

bp

76-80 °C/11 mmHg (lit.)

density

1.437 g/mL at 25 °C (lit.)

functional group

fluoro
nitro

SMILES string

[O-][N+](=O)c1ccc(F)c(F)c1

InChI

1S/C6H3F2NO2/c7-5-2-1-4(9(10)11)3-6(5)8/h1-3H

InChI key

RUBQQRMAWLSCCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The experimental and computational thermochemical study of 3,4-difluoronitrobenzene was studied[1].

Application

3,4-Difluoronitrobenzene was used in the preparation of xanthones and acridones[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oludotun A Phillips et al.
Journal of enzyme inhibition and medicinal chemistry, 35(1), 1471-1482 (2020-07-09)
Oxazolidinone hydroxamic acid derivatives were synthesised and evaluated for inhibitory activity against leukotriene (LT) biosynthesis in three in vitro cell-based test systems and on direct inhibition of recombinant human 5-lipoxygenase (5-LO). Thirteen of the 19 compounds synthesised were considered active ((50%
Yumiko Suzuki et al.
Chemical & pharmaceutical bulletin, 54(12), 1653-1658 (2006-12-02)
Xanthones and acridones were synthesized from 3,4-difluoronitrobenzene and 2-fluorobenzaldehydes in two or three steps. The key step was nucleophilic aroylation catalyzed by imidazolidenyl carbene. The nucleophilic aroylation of 3,4-difluoronitrobenzene afforded 2,2'-difluoro-4-nitrobenzophenones. The cyclization of the difluorobenzophenones with O-nucleophile and N-nucleophile
Manuel A V Ribeiro da Silva et al.
The journal of physical chemistry. B, 114(40), 12914-12925 (2010-09-24)
This work reports the experimental and computational thermochemical study performed on three difluorinated nitrobenzene isomers: 2,4-difluoronitrobenzene (2,4-DFNB), 2,5-difluoronitrobenzene (2,5-DFNB), and 3,4-difluoronitrobenzene (3,4-DFNB). The standard (p° = 0.1 MPa) molar enthalpies of formation in the liquid phase of these compounds were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service