Skip to Content
Merck
All Photos(1)

Key Documents

261793

Sigma-Aldrich

3-Benzoylbenzoic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COC6H4CO2H
CAS Number:
Molecular Weight:
226.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

powder

mp

164-166 °C (lit.)

functional group

carboxylic acid
ketone
phenyl

SMILES string

OC(=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C14H10O3/c15-13(10-5-2-1-3-6-10)11-7-4-8-12(9-11)14(16)17/h1-9H,(H,16,17)

InChI key

AXJXRLHTQQONQR-UHFFFAOYSA-N

Application

3-Benzoylbenzoic acid has been used in:
  • light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces[1]
  • preparation of benzophenone flavonol derivative[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of flavonol derivatives as probes of biological processes.
Tanaka H, et al.
Tetrahedron Letters, 41(50), 9735-9739 (2000)
Sabine Szunerits et al.
Chemical communications (Cambridge, England), (27)(27), 2793-2795 (2007-07-05)
Irradiation of a patterned benzophenone-terminated boron-doped diamond (BDD) surface with UV light (lambda = 350 nm) in the presence of a 15(mer) oligonucleotide resulted in the covalent linking of the DNA strand to the BDD interface.
Marta T Ignasiak et al.
The journal of physical chemistry. B, 118(29), 8549-8558 (2014-06-20)
The Met residue oxidation has been studied for decades. Although many efforts have been made on the identification of free radicals, some doubts remain about their final fates, i.e., the nature of stable oxidation products. The photosensitized oxidation processes of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service