Skip to Content
Merck
All Photos(1)

Key Documents

254320

Sigma-Aldrich

Zinc telluride

−100 mesh, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$543.00

$543.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$543.00

About This Item

Linear Formula:
ZnTe
CAS Number:
Molecular Weight:
192.99
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

$543.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

deposition grade

Assay

99.99% trace metals basis

form

powder

particle size

−100 mesh

density

6.34 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Zn]=[Te]

InChI

1S/Te.Zn

InChI key

NSRBDSZKIKAZHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service