Skip to Content
Merck
All Photos(1)

Key Documents

261262

Sigma-Aldrich

Scandium

powder, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sc
CAS Number:
Molecular Weight:
44.96
EC Number:
MDL number:
UNSPSC Code:
12141613
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

powder

resistivity

50.5 μΩ-cm, 0°C

bp

2836 °C (lit.)

mp

1540 °C (lit.)

density

2.99 g/mL at 25 °C (lit.)

SMILES string

[Sc]

InChI

1S/Sc

InChI key

SIXSYDAISGFNSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in solid state synthesis of unusual clusters, Sc19Br28Z4, (Z=Mn, Fe, Ru or Os). These clusters are of interest for their structure and magnetic properties.[1]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Olmos et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(16), 4894-4901 (2012-03-15)
This study demonstrates the first zeolite-catalyzed synthesis of piperidine derivatives, including peptidomimetics and indoloquinolizidine alkaloids. The approach developed utilizes a highly effective one-pot reaction cascade, through imine formation and imino-Diels-Alder reactions, promoted by scandium-loaded zeolites as a heterogeneous catalyst. The
Jacob S Kanady et al.
Journal of the American Chemical Society, 135(3), 1073-1082 (2012-12-18)
The oxygen-evolving complex (OEC) of photosystem II contains a Mn(4)CaO(n) catalytic site, in which reactivity of bridging oxidos is fundamental to OEC function. We synthesized structurally relevant cuboidal Mn(3)MO(n) complexes (M = Mn, Ca, Sc; n = 3,4) to enable
Asymmetric synthesis of β-amino nitriles through a Sc(III)-catalyzed three-component Mannich reaction of silyl ketene imines.
Jiannan Zhao et al.
Angewandte Chemie (International ed. in English), 52(12), 3473-3477 (2013-02-14)
Wei Li et al.
Angewandte Chemie (International ed. in English), 51(34), 8644-8647 (2012-07-24)
Asymmetric expansion: A catalytic asymmetric ring-expansion reaction of the title compounds occurs in the presence of a Sc(OTf)(3) catalyst bearing an N,N'-dioxide-based ligand. Highly functionalized 2-quinolone derivatives containing a chiral C4-quaternary stereocenter were obtained in high yields and high levels
Hiroshi Inaba et al.
Dalton transactions (Cambridge, England : 2003), 41(37), 11424-11427 (2012-08-15)
We have succeeded in preparing semi-synthesized proteins bound to Sc(3+) ion which can promote an epoxide ring-opening reaction. The Sc(3+) binding site was created on the surface of [(gp5βf)(3)](2) (N. Yokoi et al., Small, 2010, 6, 1873) by introducing a

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service