Skip to Content
Merck
All Photos(2)

Documents

GF31290207

Scandium

foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%

Synonym(s):

Scandium, SC000200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sc
CAS Number:
Molecular Weight:
44.96
MDL number:
UNSPSC Code:
12141613
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

foil

manufacturer/tradename

Goodfellow 312-902-07

resistivity

50.5 μΩ-cm, 0°C

size × thickness

25x25 mm × 0.025 mm

bp

2836 °C (lit.)

mp

1540 °C (lit.)

density

2.99 g/mL at 25 °C (lit.)

SMILES string

[Sc]

InChI

1S/Sc

InChI key

SIXSYDAISGFNSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Duchemin et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 103, 160-165 (2015-06-24)
Experimental cross sections for deuteron induced nuclear reactions on natural titanium were measured, using the stacked-foil technique and gamma spectrometry, up to 34 MeV with beams provided by the ARRONAX cyclotron. The experimental cross section values were monitored using the
Cristina Müller et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(10), 1658-1664 (2014-07-19)
In recent years, (47)Sc has attracted attention because of its favorable decay characteristics (half-life, 3.35 d; average energy, 162 keV; Eγ, 159 keV) for therapeutic application and for SPECT imaging. The aim of the present study was to investigate the
Jasmin Lother et al.
International journal of medical microbiology : IJMM, 304(8), 1160-1168 (2014-09-10)
The mould Aspergillus fumigatus is primarily an opportunistic pathogen of immunocompromised patients. Once fungal spores have been inhaled they encounter cells of the innate immune system, which include dendritic cells (DCs). DCs are the key antigen-presenting cells of the immune
Heleen Vanneste et al.
Scientific reports, 5, 11670-11670 (2015-07-02)
Atmospheric dust loadings play a crucial role in the global climate system. Southern South America is a key dust source, however, dust deposition rates remain poorly quantified since the last glacial termination (~17 kyr ago), an important timeframe to anticipate
Lu Qin et al.
PloS one, 10(4), e0124887-e0124887 (2015-05-01)
The BolA-like protein is present in all eukaryotes, and it is able to form complex with monothiol glutaredoxin of the same subcellular compartments, suggesting that the BolA-like protein has essential function in eukaryotes, and that the function is associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service