Skip to Content
Merck
All Photos(1)

Key Documents

257168

Sigma-Aldrich

Triethylaluminum

93%

Synonym(s):

Aluminumtriethanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(C2H5)3
CAS Number:
Molecular Weight:
114.16
Beilstein:
3587229
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 62.2 °C)

Quality Level

Assay

93%

bp

128-130 °C/50 mmHg (lit.)

mp

−50 °C (lit.)

density

0.835 g/mL at 25 °C (lit.)

SMILES string

CC[Al](CC)CC

InChI

1S/3C2H5.Al/c3*1-2;/h3*1H2,2H3;

InChI key

VOITXYVAKOUIBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylaluminum can undergo 1,4-addition to 2-cyclopentenone in the presence of a chiral diphosphite ligand to form 3-ethylcyclopentanone.[1]

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Self-heat. 2 - Skin Corr. 1A - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-catalyzed enantioselective conjugate addition of triethylaluminum to 2-cyclopentenone
Su L, et al.
Tetrahedron Asymmetry, 14(13), 1865-1869 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service