Skip to Content
Merck
All Photos(3)

Key Documents

255130

Sigma-Aldrich

N-Methyl-N-vinylacetamide

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$412.00
500 ML
$1,510.00

$412.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$412.00
500 ML
$1,510.00

About This Item

Linear Formula:
CH3CON(CH3)CH=CH2
CAS Number:
Molecular Weight:
99.13
Beilstein:
1743331
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$412.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.484 (lit.)

bp

70 °C/25 mmHg (lit.)

density

0.959 g/mL at 25 °C (lit.)

SMILES string

CN(C=C)C(C)=O

InChI

1S/C5H9NO/c1-4-6(3)5(2)7/h4H,1H2,2-3H3

InChI key

PNLUGRYDUHRLOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service