Skip to Content
Merck
All Photos(1)

Key Documents

255068

Sigma-Aldrich

Tris(methylthio)methane

98%

Synonym(s):

Trimethyl trithioorthoformate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$102.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$102.00

About This Item

Linear Formula:
(CH3S)3CH
CAS Number:
Molecular Weight:
154.32
Beilstein:
1733825
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.577 (lit.)

bp

102 °C/15 mmHg (lit.)
225-227 °C (lit.)

mp

16 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

CSC(SC)SC

InChI

1S/C4H10S3/c1-5-4(6-2)7-3/h4H,1-3H3

InChI key

YFMZQCCTZUJXEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(methylthio)methane is a carboxylic anion equivalent.

Application

Tris(methylthio)methane has been used in synthesis of:
  • (−)-nephrosteranic acid and (−)-roccellaric acid[1]
  • (4R,5R)-5-({[1-(tert-butyl)-1,1-diphenylsilyl]oxy}methyl)4-[tri(methylthio)methyl]tetrahydro-furan-2-one[2]
  • terminal difluoromethylenes[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Mugnaini et al.
Bioorganic & medicinal chemistry, 11(3), 357-366 (2003-01-09)
Novel nucleoside analogues of both D and L enantiomeric series were prepared by coupling reaction between a 2',3'-dideoxy-3'-modified furanose moiety and four different nucleobases. Though in all cases anomeric mixtures of nucleosides were obtained, the presence of the sterically bulky
From ketones, aldehydes or alkyl halides to terminal 1, 1-difluoroolefins using BrF3.
Hagooly A and Rozen S.
Journal of Fluorine Chemistry, 126(8), 1239-1245 (2005)
Bin Mao et al.
Organic letters, 13(5), 948-951 (2011-01-28)
An efficient catalytic asymmetric synthesis of chiral γ-butenolides was developed based on the hetero-allylic asymmetric alkylation (h-AAA) in combination with ring closing metathesis (RCM). The synthetic potential of the h-AAA-RCM protocol was illustrated with the facile synthesis of (-)-whiskey lactone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service