Skip to Content
Merck
All Photos(2)

Key Documents

252999

Sigma-Aldrich

Nialamide

95%

Synonym(s):

N-Benzyl-β-(isonicotinylhydrazino)propionamide, N-Isonicotinoyl-N′-[β-(N-benzylcarboxamido)ethyl]hydrazine, Pyridine-4-carboxylic 2-[2-(benzylcarbamoyl)ethyl]hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O2
CAS Number:
Molecular Weight:
298.34
Beilstein:
492941
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

152-154 °C (lit.)

solubility

methanol: soluble 10 mg/mL, clear, colorless to faintly yellow

functional group

amide
amine
hydrazide
phenyl

SMILES string

O=C(CCNNC(=O)c1ccncc1)NCc2ccccc2

InChI

1S/C16H18N4O2/c21-15(18-12-13-4-2-1-3-5-13)8-11-19-20-16(22)14-6-9-17-10-7-14/h1-7,9-10,19H,8,11-12H2,(H,18,21)(H,20,22)

InChI key

NOIIUHRQUVNIDD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Nialamide is an antidepressant drug and its effect on β-adrenergic receptor-adenylate cyclase system of rat pineal gland has been studied[1]. Antidepressant action of the combination of nialamide and 5-hydroxytryptophan has been evaluated[2].

Biochem/physiol Actions

Non-selective MAO-A/B inhibitor.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C F Meehan et al.
The Journal of physiology, 590(2), 289-300 (2011-11-23)
Recently, transgenic mice have been created with mutations affecting the components of the mammalian spinal central pattern generator (CPG) for locomotion; however, it has currently only been possible to evoke fictive locomotion in mice, using neonatal in vitro preparations. Here, we demonstrate
F Datiche et al.
Brain research, 671(1), 27-37 (1995-02-06)
Retrograde axonal transport of the cholera toxin B subunit (CTb) was combined with 5-HT immunohistochemistry to determine the origin of the serotonergic innervation of the piriform cortex (PC) in the rat. After iontophoretic CTb injections in the PC, a substantial
Subsensitivity of the beta-adrenergic receptor-linked adenylate cyclase system of rat pineal gland following repeated treatment with desmethylimipramine and nialamide.
J A Moyer et al.
Molecular pharmacology, 19(2), 187-193 (1981-03-01)
P J Whelan et al.
Journal of neurophysiology, 78(3), 1643-1650 (1997-10-06)
Previous studies have shown that stimulation of group 'I' afferents from ankle extensor muscles can prolong the cycle period in decerebrate walking cats and that the magnitude of these effects can be altered after chronic axotomy of the lateral-gastrocnemius/soleus (LGS)
J J Aliño et al.
International pharmacopsychiatry, 11(1), 8-15 (1976-01-01)
Through a controlled double-blind study in 30 hospitalized patients affected with endogenous depression, the antidepressant action of the combination of nialamide+l-5-HTP has been evaluated and compared with a control group which only received nialamide (+ placebo). The patients treated with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service