Skip to Content
Merck
All Photos(1)

Key Documents

251089

Sigma-Aldrich

2-(Methylthio)pyridine-3-carboxylic acid

98%

Synonym(s):

2-(Methylthio)nicotinic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$318.00

$318.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$318.00

About This Item

Empirical Formula (Hill Notation):
C7H7NO2S
CAS Number:
Molecular Weight:
169.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$318.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

98%

form

solid

mp

215-217 °C (lit.)

functional group

carboxylic acid
thioether

SMILES string

CSc1ncccc1C(O)=O

InChI

1S/C7H7NO2S/c1-11-6-5(7(9)10)3-2-4-8-6/h2-4H,1H3,(H,9,10)

InChI key

COPSJQVPEUUOKY-UHFFFAOYSA-N

General description

Anisotropy in 2-(methylthio)pyridine-3-carboxylic acid [2-(methylthio)nicotinic acid] has been investigated by nanoindentation experiment[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Malla Reddy et al.
Chemical communications (Cambridge, England), (31)(31), 3945-3947 (2005-08-03)
Bending is observed in organic crystals when the packing is anisotropic in such a way that strong and weak interaction patterns occur in nearly perpendicular directions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service