Skip to Content
Merck
All Photos(1)

Documents

249769

Sigma-Aldrich

2-Ethylhexyl chloroformate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
192.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

>1 (vs air)

vapor pressure

0.1 psi ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.431 (lit.)

bp

106-107 °C/30 mmHg (lit.)

density

0.981 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCC(CC)COC(Cl)=O

InChI

1S/C9H17ClO2/c1-3-5-6-8(4-2)7-12-9(10)11/h8H,3-7H2,1-2H3

InChI key

RTGLJCSUKOLTEM-UHFFFAOYSA-N

Application

2-Ethylhexyl chloroformate has been used in the synthesis of:
  • 1,2,4-oxadiazol-5-ones
  • 3-(4-fluorophenyl)-1,2,4-oxadiazol-5(4H)-one
  • (alkoxycarbonyloxy)carbamimidoyl derivatives

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding interactions between 3-aryl-1, 2, 4-oxadiazol-5-ones and a trisimidazoline base.
Reichert A, et al.
Journal of the Chemical Society. Perkin Transactions 1, 11, 1321-1328 (2001)
Month 2013 Synthesis of Azilsartan and Its Selected Potential Impurities.
Radl S, et al.
Journal of Heterocyclic Chemistry, 50(4), 929-936 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service