Skip to Content
Merck
All Photos(1)

Key Documents

252778

Sigma-Aldrich

Hexyl chloroformate

97%

Synonym(s):

Chloroformic acid n-hexyl ester, Hexyl carbonochloridate, Hexyl chlorocarbonate, Hexyloxycarbonyl chloride, n-Hexyl chloroformate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$75.90
25 G
$336.00

$75.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$75.90
25 G
$336.00

About This Item

Linear Formula:
ClCO2(CH2)5CH3
CAS Number:
Molecular Weight:
164.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

0.06 psi ( 20 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.424 (lit.)

bp

60-61 °C/7 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

CCCCCCOC(Cl)=O

InChI

1S/C7H13ClO2/c1-2-3-4-5-6-10-7(8)9/h2-6H2,1H3

InChI key

KIWBRXCOTCXSSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexyl chloroformate has been used as derivatization reagent in:
  • determination of sarcosine and N-ethylglycine in urine and urinary sediments by solid-phase micro extraction and fast GC-MS analysis[1]
  • determination of benzoylecgonine in urine by GC-quadrupole ion trap mass spectrometry[2]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B J Hall et al.
Journal of forensic sciences, 44(3), 527-534 (1999-07-17)
A derivatization/solid phase microextraction (SPME) method for the determination of benzoylecgonine in urine was developed. The derivatization is conducted directly in 1 mL of urine while sonicating for 3 min with 12 microL of hexyl chloroformate and 70 microL of
Tomáš Hložek et al.
Journal of analytical toxicology, 40(9), 749-753 (2016-09-04)
A simple, sensitive and robust method for simultaneous determination of antiepileptic drugs (gabapentin, pregabalin and vigabatrin) in human serum using GC-MS was developed and validated for clinical toxicology purposes. This method employs an emerging class of derivatization agents - alkyl
F Bianchi et al.
Analytica chimica acta, 707(1-2), 197-203 (2011-10-27)
A fully automated, non invasive, rapid and high-throughput method for the direct determination of sarcosine and N-ethylglycine in urine and urinary sediments using hexyl chloroformate derivatization followed by direct immersion solid-phase micro extraction and fast gas chromatography-mass spectrometric analysis was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service