Skip to Content
Merck
All Photos(1)

Key Documents

241180

Sigma-Aldrich

2-Chlorobenzyl chloride

≥99%

Synonym(s):

α,2-Dichlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2Cl
CAS Number:
Molecular Weight:
161.03
Beilstein:
471700
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.5 (vs air)

vapor pressure

3 mmHg ( 84 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.559 (lit.)

bp

213-214 °C (lit.)

density

1.274 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClCc1ccccc1Cl

InChI

1S/C7H6Cl2/c8-5-6-3-1-2-4-7(6)9/h1-4H,5H2

InChI key

BASMANVIUSSIIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Alarie
Archives of toxicology, 72(5), 277-282 (1998-06-18)
We expanded a previously described rule-based computerized method to recognize the sensory irritating effect of airborne chemicals. Using 2-chlorobenzylchloride (CBC) as a sensory irritant, characteristic modifications of the normal breathing pattern of exposed mice were evaluated by measuring the duration
N Ariga et al.
Journal of biochemistry, 88(1), 97-102 (1980-07-01)
o-Chlorobenzylchloride, a simple aromatic halogen compound, was found to inhibit the growth of Saccharomyces cerevisiae and to lower the contents of sterols and fatty acids. The growth inhibition was considerably alleviated by the presence of sterols such as ergosterol and
R Vijayaraghavan et al.
Archives of toxicology, 68(8), 490-499 (1994-01-01)
The pattern and timing of a normal breath in unanesthetized mice was analyzed from measurement of inspiratory and expiratory airflows (VI and VE). Airflow was measured via a differential pressure transducer, attached to a pneumotachograph, which itself was attached to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service