Skip to Content
Merck
All Photos(1)

Key Documents

238023

Sigma-Aldrich

Lithium sulfate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2SO4
CAS Number:
Molecular Weight:
109.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
99%
form:
solid

Assay

99%

form

solid

mp

845 °C (lit.)

density

2.22 g/mL at 25 °C (lit.)

SMILES string

[Li+].[Li+].[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

INHCSSUBVCNVSK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L K Singh et al.
The National medical journal of India, 24(3), 151-152 (2011-07-27)
Lithium-induced cardiotoxicity, though rare at therapeutic levels, has been reported frequently in overdoses. We report a patient who developed sinus bradycardia while being treated with lithium carbonate even though the serum lithium levels were within the therapeutic range. It reversed
Benedikt Habermeyer et al.
Psychiatrische Praxis, 35(4), 198-200 (2007-11-08)
Lithium is known to be neurotoxic at higher serum levels. In rare cases patients develop symptoms of intoxication or full blown intoxications even when lithium levels are normal. Our aim was to detect predicting factors of lithium neurotoxicity at normal
Sanjay Kasture et al.
Natural product research, 25(20), 1950-1954 (2011-06-29)
In the traditional system of medicine in Sardinia, Rubia peregrina (RP) is reported as an aphrodisiac herb. Since aphrodisiacs may also have dopaminergic activity, and there can be a reciprocal relationship between dopaminergic and serotonergic functions in the central nervous
Yuya Yamada et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 62(Pt 10), 1016-1017 (2006-10-03)
Atg3 is an E2-like enzyme that catalyzes the conjugation reaction between Atg8 and phosphatidylethanolamine (PE). The Atg8-PE conjugate is essential for autophagy, the bulk degradation process of cytoplasmic components by the vacuolar/lysosomal system. Crystals of Saccharomyces cerevisiae Atg3 have been
Janaína Teles de Faria et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(27), 3031-3037 (2009-09-01)
Xylose reductase (XR) from Debaryomyces hansenii was extracted by partitioning in aqueous two-phase systems (ATPS) composed of polyethylene glycol (PEG) 4000 in the presence of different salts, specifically sodium sulfate, lithium sulfate and potassium phosphate. Batch extractions were carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service