Skip to Content
Merck
All Photos(1)

Documents

233196

Sigma-Aldrich

2,6-Bis(trifluoromethyl)benzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3CO2H
CAS Number:
Molecular Weight:
258.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

138-140 °C (lit.)

SMILES string

OC(=O)c1c(cccc1C(F)(F)F)C(F)(F)F

InChI

1S/C9H4F6O2/c10-8(11,12)4-2-1-3-5(9(13,14)15)6(4)7(16)17/h1-3H,(H,16,17)

InChI key

XZNLSDPNMNWCRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Bis(trifluoromethyl)benzoic acid has been used in selective separation of brominated aromatic compounds using the polymer-based separation media by the fragment imprinting technique.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective separation of brominated bisphenol A homologues using a polymer-based medium prepared by the fragment imprinting technique.
Kubo T, et al.
Analytica Chimica Acta, 549(1), 45-50 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service