Skip to Content
Merck
All Photos(1)

Documents

23070

Sigma-Aldrich

(1-Chloroethyl)trimethylsilane

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13ClSi
CAS Number:
Molecular Weight:
136.70
Beilstein:
1732094
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.422

bp

116 °C (lit.)

density

0.864 g/mL at 20 °C (lit.)

SMILES string

CC(Cl)[Si](C)(C)C

InChI

1S/C5H13ClSi/c1-5(6)7(2,3)4/h5H,1-4H3

InChI key

UWJVDEZZIPJQRF-UHFFFAOYSA-N

Other Notes

Treating this chlorosilane with sec-BuLi gives 1-chloro-1-trimethylsilylethyl lithium which reacts with aldehydes and ketones to form methyl ketones via an intermediate α,β-epoxysilane.; Used in an efficient synthesis of R(+)-frontalin; The Grignard reagent is useful for many reactions

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Magnus et al.
Journal of the Chemical Society. Chemical Communications, 297-297 (1978)
F. Cooke et al.
Organometallics, 1, 893-893 (1982)
J.A. Soderquist et al.
The Journal of Organic Chemistry, 45, 3571-3571 (1980)
S. Sato et al.
Journal of Organometallic Chemistry, 344, 71-71 (1988)
T. Hayashi et al.
The Journal of Organic Chemistry, 51, 3772-3772 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service