Skip to Content
Merck
All Photos(1)

Key Documents

228001

Sigma-Aldrich

3-Methyl-2-oxazolidinone

99.5%

Synonym(s):

3-Methyl-1,3-oxazolidin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$463.00

$463.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$463.00

About This Item

Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$463.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.5%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

87-90 °C/1 mmHg (lit.)

mp

15 °C (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

CN1CCOC1=O

InChI

1S/C4H7NO2/c1-5-2-3-7-4(5)6/h2-3H2,1H3

InChI key

VWIIJDNADIEEDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methyl-2-oxazolidinone mixed with ethylene carbonate or dimethyl carbonate in the presence of lithium tetrafluoroborate or lithium hexafluorophosphate has been used as an electrolyte in lithium batteries[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Is 3-methyl-2-oxazolidinone a suitable solvent for lithium-ion batteries?
Gzara L, et al.
Journal of Power Sources, 156(2), 634-644 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service