Skip to Content
Merck
All Photos(1)

Key Documents

M9009

Sigma-Aldrich

Methoxyacetonitrile

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$130.00
25 G
$579.00
100 G
$1,270.00

$130.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$130.00
25 G
$579.00
100 G
$1,270.00

About This Item

Linear Formula:
CH3OCH2CN
CAS Number:
Molecular Weight:
71.08
Beilstein:
1738014
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$130.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Assay

≥98%

form

liquid

refractive index

n20/D 1.381 (lit.)

bp

118-119 °C/731 mmHg (lit.)

density

0.956 g/mL at 25 °C (lit.)

SMILES string

COCC#N

InChI

1S/C3H5NO/c1-5-3-2-4/h3H2,1H3

InChI key

QKPVEISEHYYHRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masazumi Tamura et al.
Nature communications, 6, 8580-8580 (2015-10-06)
Multidentate materials formed by simply mixing heterogeneous and homogeneous components are promising for construction of versatile active sites on the surface of heterogeneous compounds, however, to the best of our knowledge, there are no reports on such materials. Self-assembly of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service