Skip to Content
Merck
All Photos(2)

Key Documents

226858

Sigma-Aldrich

Silver(I) fluoride

≥99.9% trace metals basis

Synonym(s):

Argentous fluoride, Silver monofluoride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$60.60
5 G
$175.00
25 G
$648.00

$60.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$60.60
5 G
$175.00
25 G
$648.00

About This Item

Empirical Formula (Hill Notation):
AgF
CAS Number:
Molecular Weight:
126.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$60.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1000.0 ppm Trace Metal Analysis

mp

≥300 °C (lit.)

density

5.852 g/mL at 25 °C (lit.)

SMILES string

F[Ag]

InChI

1S/Ag.FH/h;1H/q+1;/p-1

InChI key

REYHXKZHIMGNSE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Preparation of bromoacetylenes from bulky, trialkylsilylacetylenes.[1] Also used in a palladium-catalyzed cross coupling of acetanilides with trialkoxysilanes.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 62, 4081-4081 (2006)
Ortho arylation of acetanilides via Pd(II)-catalyzed C-H functionalization.
Shangdong Yang et al.
Journal of the American Chemical Society, 129(19), 6066-6067 (2007-04-26)

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service