Skip to Content
Merck
All Photos(1)

Key Documents

221074

Sigma-Aldrich

cis-2,4,5-Trimethoxy-1-propenylbenzene

70%

Synonym(s):

β-Asarone, cis-1-Propenyl-2,4,5-trimethoxybenzene, cis-Asarone, cis-Isoasarone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH=CHCH3
CAS Number:
Molecular Weight:
208.25
Beilstein:
1910605
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

70%

form

liquid

refractive index

n20/D 1.558 (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C)=C(/[H])c1cc(OC)c(OC)cc1OC

InChI

1S/C12H16O3/c1-5-6-9-7-11(14-3)12(15-4)8-10(9)13-2/h5-8H,1-4H3/b6-5-

InChI key

RKFAZBXYICVSKP-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-2,4,5-Trimethoxy-1-propenylbenzene is an antibiotic and was isolated from the extract of Acorus gramineus using various chromatographic procedures[1].

Application

cis-2,4,5-Trimethoxy-1-propenylbenzene (β-Asarone) was used in preparation of:
  • 2,4,5-trimethoxycinnamaldehyde and 2,4,5-trimethoxycinnamyltosylhydrazone[2]
  • 1-(2,4,5-trimethoxyphenyl)-1,2-dihydroxypropane[3]

Other Notes

Remainder trans (α-asarone)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arun K Sinha et al.
Journal of natural products, 65(5), 764-765 (2002-05-25)
Oxidation of beta-asarone (2) with DDQ gave trans-2,4,5-trimethoxycinnamaldehyde (3), which on treatment with p-toluenesulfonyl hydrazine provided corresponding alpha,beta-unsaturated hydrazone derivative (4). Reduction of 4 with sodium borohydride in acetic acid afforded gamma-asarone (1) in 43% yield.
A K Sinha et al.
Natural product letters, 15(6), 439-444 (2002-02-13)
1-(2,4,5-Trimethoxyphenyl)-1,2-dihydroxypropane (2), a natural phenylpropanoid occurring in Piper clusii, has been synthesized for the first time from toxic beta-asarone (1) of Acorus calamus with osmium tetroxide, while 1 with osmium tetroxide (catalytic amount) in presence of sodium metaperiodate furnished the
Jee Yeon Lee et al.
Journal of agricultural and food chemistry, 52(4), 776-780 (2004-02-19)
An antifungal substance was isolated from the extract of Acorus gramineus using various chromatographic procedures. The antibiotic was identified as beta-asarone, cis-2,4,5-trimethoxy-1-propenylbenzene, on the basis of the high-resolution EI-mass, NMR, and UV spectral data. Beta-asarone completely inhibited mycelial growth of
Daijie Wang et al.
Journal of separation science, 34(23), 3339-3343 (2011-11-01)
In this study, supercritical fluid extraction (SFE) was used to extract essential oil from Acorus tatarinowii Schott under the pressure of 25 MPa and temperature of 35°C. Two (E)- and (Z)-diastereomers of α-asarone and β-asarone were separated and purified by
Jadwiga Marczewska et al.
Acta poloniae pharmaceutica, 70(2), 349-354 (2013-04-26)
In our previous paper we examined mutagenic and genotoxic activity of 4 alpha-asarone isomers 2-5 exhibiting relatively high hypolipidemic activity. In the present paper, we examined genotoxic activity of alpha-asarone and its isomers as the ability to damage cellular DNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service