Skip to Content
Merck
All Photos(2)

Key Documents

215333

Sigma-Aldrich

4-Hydroxy-D-phenylglycine

≥98%

Synonym(s):

(2R)-2-Amino-2-(4-hydroxyphenyl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(NH2)CO2H
CAS Number:
Molecular Weight:
167.16
Beilstein:
2210998
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

optical activity

[α]23/D −158±3°, c = 1 in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

240 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](C(O)=O)c1ccc(O)cc1

InChI

1S/C8H9NO3/c9-7(8(11)12)5-1-3-6(10)4-2-5/h1-4,7,10H,9H2,(H,11,12)/t7-/m1/s1

InChI key

LJCWONGJFPCTTL-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Yu et al.
Folia microbiologica, 54(6), 509-515 (2010-02-09)
The first establishment of a homologous expression system in the host Ralstonia pickettii CGMCC1596 using the compatible broad-host-range plasmid pWB5 is described. When whole cells of the recombinant strain R. pickettii MMYY01 (CGMCC1596/pYY05) were used as the biocatalyst to transform
Reducing the burden of ischemia reperfusion: editorial to: "Effect of Chronic CPT-1 Inhibition on Myocardial Ischemia-Reperfusion Injury (I/R) in a Model of Diet-Induced Obesity" by G. Maarman et al.
Roger M Beadle
Cardiovascular drugs and therapy, 26(3), 193-194 (2012-03-15)
Xihou Yin et al.
Microbiology (Reading, England), 152(Pt 10), 2969-2983 (2006-09-29)
The biosynthetic gene cluster for the 17 aa peptide antibiotic enduracidin has been cloned and sequenced from Streptomyces fungicidicus ATCC 21013. The 84 kb gene cluster contains 25 ORFs and is located within a 116 kb genetic locus that was
M G Marina Prendes et al.
The journal of physiological sciences : JPS, 61(4), 303-312 (2011-05-07)
The effects of ischemic-postconditioning (IPOC) on functional recovery and cell viability of ischemic-reperfused hearts from fed and fasted rats were studied in relation to triacylglycerol and glycogen mobilization, ATP content, glucose-6-phosphate dehydrogenase activity and reduced/oxidized glutathione (GSH/GSSG). Oxidative damage was
Monica Prieto et al.
The Journal of organic chemistry, 72(3), 1047-1050 (2007-01-27)
Reaction conditions considered to be typical in Suzuki couplings can cause significant (up to 34% of the unwanted enantiomer) loss of optical purity in sensitive substrates such as hydroxyphenylglycine 1. This may be remedied using sodium succinate instead of sodium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service