Skip to Content
Merck
All Photos(1)

Documents

56160

Sigma-Aldrich

4-Hydroxy-L-phenylglycine

≥99.0% (NT)

Synonym(s):

(2S)-2-Amino-2-(4-hydroxyphenyl)acetic acid, Oxfenicine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO3
CAS Number:
Molecular Weight:
167.16
Beilstein:
3589845
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (NT)

form

solid

optical activity

[α]20/D +158±3°, c = 1% in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

N[C@H](C(O)=O)c1ccc(O)cc1

InChI

1S/C8H9NO3/c9-7(8(11)12)5-1-3-6(10)4-2-5/h1-4,7,10H,9H2,(H,11,12)/t7-/m0/s1

InChI key

LJCWONGJFPCTTL-ZETCQYMHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xihou Yin et al.
Microbiology (Reading, England), 152(Pt 10), 2969-2983 (2006-09-29)
The biosynthetic gene cluster for the 17 aa peptide antibiotic enduracidin has been cloned and sequenced from Streptomyces fungicidicus ATCC 21013. The 84 kb gene cluster contains 25 ORFs and is located within a 116 kb genetic locus that was
Reducing the burden of ischemia reperfusion: editorial to: "Effect of Chronic CPT-1 Inhibition on Myocardial Ischemia-Reperfusion Injury (I/R) in a Model of Diet-Induced Obesity" by G. Maarman et al.
Roger M Beadle
Cardiovascular drugs and therapy, 26(3), 193-194 (2012-03-15)
H Yu et al.
Folia microbiologica, 54(6), 509-515 (2010-02-09)
The first establishment of a homologous expression system in the host Ralstonia pickettii CGMCC1596 using the compatible broad-host-range plasmid pWB5 is described. When whole cells of the recombinant strain R. pickettii MMYY01 (CGMCC1596/pYY05) were used as the biocatalyst to transform
Isidore C Okere et al.
Clinical and experimental pharmacology & physiology, 34(1-2), 113-119 (2007-01-05)
1. Cardiac lipotoxicity is characterized by hypertrophy and contractile dysfunction and can be triggered by impaired mitochondrial fatty acid oxidation and lipid accumulation. The present study investigated the effect of dietary fatty acid intake alone and in combination with inhibition
M G Marina Prendes et al.
The journal of physiological sciences : JPS, 61(4), 303-312 (2011-05-07)
The effects of ischemic-postconditioning (IPOC) on functional recovery and cell viability of ischemic-reperfused hearts from fed and fasted rats were studied in relation to triacylglycerol and glycogen mobilization, ATP content, glucose-6-phosphate dehydrogenase activity and reduced/oxidized glutathione (GSH/GSSG). Oxidative damage was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service