Skip to Content
Merck
All Photos(1)

Key Documents

214019

Sigma-Aldrich

2-Methyl-1-pentanol

99%

Synonym(s):

(±)-2-Methyl-1-pentanol, 2-Methylpentanol-1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)CH2OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718974
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

Assay

99%

form

liquid

expl. lim.

9.6 %

refractive index

n20/D 1.418 (lit.)

bp

148 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCC(C)CO

InChI

1S/C6H14O/c1-3-4-6(2)5-7/h6-7H,3-5H2,1-2H3

InChI key

PFNHSEQQEPMLNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonaqueous lipase-catalyzed kinetic resolution of racemic 2-methyl-1-pentanol in a continuously operated fixed bed reactor has been reported[1]. Molar excess enthalpies of binary mixtures of 2-methyl-1-pentanol with n-hexane and its isomers has been measured in a flow microcalorimeter[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Indlekofer et al.
Biotechnology and bioengineering, 52(4), 459-471 (1996-11-20)
The nonaqueous lipase-catalyzed kinetic resolution of racemic 2-methyl-1-pentanol in a continuously operated fixed bed reactor was theoretically and experimentally studied. A 2-dimensional mathematical model was developed to predict the performance of the heterogeneous enantioselective biotransformation and to optimize the productivity
Excess enthalpies of binary mixtures of 2-methyl-1-pentanol with hexane isomers.
Kimura F and Benson GC.
Fluid Phase Equilibria, 8(1), 107-112 (1982)
Jung-Min Hong et al.
Foods (Basel, Switzerland), 9(9) (2020-09-25)
Volatile compositions and sensory characteristics of 11 commercially distilled soju samples were investigated using headspace solid-phase microextraction (HS-SPME) with gas chromatography-mass spectrometry (GC-MS) and sensory descriptive analysis. A total of 59 major volatile compounds, consisting of 32 esters, 10 alcohols
Kathrin Vollmer et al.
Food chemistry, 345, 128786-128786 (2020-12-15)
The effects of single and twofold continuous pressure change technology (PCT) applications on the volatiles and sensory characteristics of pineapple juice were studied. Fresh and thermally pasteurised juices were additionally characterised. 128 volatiles were tentatively assigned in the four juices.
Neepa Choksi et al.
Cutaneous and ocular toxicology, 39(3), 180-192 (2020-06-27)
OptiSafe is an in chemico test method that identifies potential eye irritants based on macromolecular damage following test chemical exposure. The OptiSafe protocol includes a prescreen assessment that identifies test chemicals that are outside the applicability domain of the test

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service