Skip to Content
Merck
All Photos(2)

Key Documents

111120

Sigma-Aldrich

2-Methyl-2-pentanol

99%

Synonym(s):

Dimethyl propyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C(CH3)2OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718951
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Quality Level

Assay

99%

refractive index

n20/D 1.411 (lit.)

bp

120-122 °C (lit.)

mp

−109-−107 °C (lit.)

density

0.835 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCC(C)(C)O

InChI

1S/C6H14O/c1-4-5-6(2,3)7/h7H,4-5H2,1-3H3

InChI key

WFRBDWRZVBPBDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-2-pentanol is a metabolite found in the urine of rats exposed to methylpentane.

Application

2-Methyl-2-pentanol has been used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michael F Wilkemeyer et al.
Molecular pharmacology, 62(5), 1053-1060 (2002-10-23)
1-Octanol antagonizes ethanol inhibition of L1-mediated cell adhesion and prevents ethanol teratogenesis in mouse whole embryo culture. Herein, we identify a new series of alcohol antagonists and study their mechanism of action. Cell aggregation assays were carried out in ethanol-sensitive
T Kawai et al.
Occupational and environmental medicine, 52(11), 757-763 (1995-11-01)
To investigate the possibilities of personal ambient monitoring and biological monitoring for methylpentane isomers. The performance of activated carbon cloth to absorb 2- and 3-methylpentane was studied by experimental vapour exposure followed by solvent extraction and gas chromatography (GC). Urine
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service