Skip to Content
Merck
All Photos(3)

Documents

213209

Sigma-Aldrich

Lithium citrate hydrate

97%

Synonym(s):

Citric acid trilithium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiO2CCH2C(OH)(CO2Li)CH2CO2Li · xH2O
CAS Number:
Molecular Weight:
209.92 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

112 °C (dec.) (lit.)

solubility

water: soluble 10%, clear, colorless

SMILES string

[Li+].[Li+].[Li+].O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Li.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;1H2/q;3*+1;/p-3

InChI key

OSJRPECIECUBOG-UHFFFAOYSA-K

Application

Lithium citrate was used in the preparation of Portland cement plates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaotropic substances and their effects on the mechanical strength of Portland cement-based materials.
Rossetto HL, et al.
Materials Research Bulletin, 11(2), 183-185 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service