Skip to Content
Merck
All Photos(1)

Key Documents

1.07835

Supelco

Antimony(III) oxide

EMPLURA®

Synonym(s):

Antimony(III) oxide, di-Antimony trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O3Sb2
CAS Number:
Molecular Weight:
291.52
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-175-0
NACRES:
NA.21
Assay:
≥99.0% (bromometric)
form:
solid
solubility:
2.70 mg/L
Pricing and availability is not currently available.

vapor pressure

13.3 hPa ( 660 °C)

Quality Level

product line

EMPLURA®

Assay

≥99.0% (bromometric)

form

solid

potency

>34600 mg/kg LD50, oral (Rat)

impurities

≤1% Matter insoluble in tartaric acid

particle size

1-3 μm (D 50)

bp

1550 °C/1013 hPa

mp

656 °C (sublimed)

solubility

2.70 mg/L

density

5.4 g/cm3

bulk density

800‑1300 kg/m3

cation traces

Al: ≤20 ppm
As: ≤500 ppm
Ca: ≤20 ppm
Cu: ≤20 ppm
Fe: ≤50 ppm
K: ≤10 ppm
Mg: ≤20 ppm
Na: ≤20 ppm
Pb: ≤500 ppm

storage temp.

2-30°C

SMILES string

[Sb+3].[Sb+3].[O-2].[O-2].[O-2]

InChI

1S/3O.2Sb/q3*-2;2*+3

InChI key

GHPGOEFPKIHBNM-UHFFFAOYSA-N

Related Categories

Application


  • Kirkwood-Buff Analysis of Binary and Ternary Systems Consisting of Alcohols (Methanol, Ethanol, 1-Propanol, and 2-Propanol), Water, and n-Hexane to Understand the Formation of Surfactant-Free Microemulsions.: This research elucidates the molecular interactions in microemulsions involving 1-Propanol, offering insights into its use in creating stable, surfactant-free systems (Seo et al., 2024).



Analysis Note

Assay (bromometric): ≥ 99.0 %
Matter insoluble in tartaric acid: ≤ 1.0 %
Al (Aluminium): ≤ 20 ppm
As (Arsenic): ≤ 500 ppm
Ca (Calcium): ≤ 20 ppm
Cu (Copper): ≤ 20 ppm
Fe (Iron): ≤ 50 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 20 ppm
Na (Sodium): ≤ 20 ppm
Pb (Lead): ≤ 500 ppm
Particle size (D 50): 1.0 - 3.0 µm

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service