Skip to Content
Merck
All Photos(2)

Documents

210439

Sigma-Aldrich

Acid Blue 113

Dye content 50 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H21N5Na2O6S2
CAS Number:
Molecular Weight:
681.65
Colour Index Number:
26360
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 50%

λmax

566 nm

ε (extinction coefficient)

≥18000 at 563-569 nm in H2O at 0.02 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1cccc(c1)\N=N\c2ccc(\N=N\c3ccc(Nc4ccccc4)c5c(cccc35)S([O-])(=O)=O)c6ccccc26

InChI

1S/C32H23N5O6S2.2Na/c38-44(39,40)23-11-6-10-22(20-23)34-35-27-16-17-28(25-13-5-4-12-24(25)27)36-37-29-18-19-30(33-21-8-2-1-3-9-21)32-26(29)14-7-15-31(32)45(41,42)43;;/h1-20,33H,(H,38,39,40)(H,41,42,43);;/q;2*+1/p-2/b35-34+,37-36+;;

InChI key

XPRMZBUQQMPKCR-XGQONITOSA-L

General description

Acid Blue 113 is a diazo dye widely used in the textile industry and tannery. The dye has been used in adsorption and degradation studies to decolorize and mineralize the toxic intermediates like aromatic amines produced in the textile effluent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E van Pelt-Verkuil
Microscopica acta, 86(4), 323-338 (1982-11-01)
Apart from the applicability of the tannic acid-phosphomolybdic acid-Levanol-Fast Cyanine 5RN method for the identification of muscle myosins in Carnoy-fixed tissues, this technique can also be applied on tissue sections fixed with formalin or carbodiimide. The use of carbodiimide as
Sudharshan Sekar et al.
Biotechnology progress, 28(6), 1400-1408 (2012-08-29)
Bioreaction calorimetric studies of degradation of the dye acid blue 113 by Staphylococcus lentus are reported for the first time. The heat released during the dye degradation process can be successfully measured using reaction calorimeter. Power time and oxygen uptake
Fei Lu et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 84(1), 1-8 (2006-02-08)
A supramolecular complex of bovine serum albumin (BSA) with acid cyanine 5R (AC 5R, C.I. acid blue 113, C.I.: 26360) has been shown to form in Tris-HCl buffer solution (pH 7.42) by linear sweep voltammetry (LSV), fluorimetry, and spectrophotometry. The
V K Gupta et al.
Journal of hazardous materials, 186(1), 891-901 (2010-12-18)
A mesoporous carbon developed from waste tire rubber, characterized by chemical analysis, FTIR, and SEM studies, was used as an adsorbent for the removal and recovery of a hazardous azo dye, Acid Blue 113. Surface area, porosity, and density were
Ghanem Zayani et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 43(2), 202-209 (2008-01-04)
The apparent first-order rate constant, k(app), of the Acid Blue 113 textile azo dye photocatalytic disappearance in aqueous suspensions of 4 commercialized TiO(2) samples (50 m(2) g(-1)-Degussa P25; 54 m(2) g(-1)-Millennium Chemicals PC50; 250-270 m(2) g(-1)-Sachtleben Chemie Hombikat UV100; 320

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service