Skip to Content
Merck
All Photos(2)

Key Documents

49823

Supelco

Acid Red 73

analytical standard

Synonym(s):

Brilliant Crocein MOO, 7-Hydroxy-8-(4-phenylazophenylazo)-1,3-naphthalenedisulfonic acid disodium salt, Acid Red 73, Brilliant crocein scarlet MOO, Crocein Scarlet MOO

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$190.00

$190.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$190.00

About This Item

Empirical Formula (Hill Notation):
C22H14N4Na2O7S2
CAS Number:
Molecular Weight:
556.48
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$190.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].Oc1ccc2cc(cc(c2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O7S2.2Na/c27-19-11-6-14-12-18(34(28,29)30)13-20(35(31,32)33)21(14)22(19)26-25-17-9-7-16(8-10-17)24-23-15-4-2-1-3-5-15;;/h1-13,27H,(H,28,29,30)(H,31,32,33);;/q;2*+1/p-2/b24-23+,26-25+;;

InChI key

PEAGNRWWSMMRPZ-RVTJCSDESA-L

Looking for similar products? Visit Product Comparison Guide

General description

Acid Red 73 is a synthetically prepared sulfonated azo dye.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Adsorption of acid dyes on chitosan-equilibrium isotherm analyses.
Wong CY, et al.
Process Biochemistry (Oxford, United Kingdom), 39(6), 695-704 (2004)
The analysis of some sulphonated azo dyes by mixed mode HPLC.
Weatherall LI.
Journal of Liquid Chromatography, 14(10), 1903-1912 (1991)
Yuezhong Wen et al.
Journal of hazardous materials, 201-202, 162-169 (2011-12-16)
There is a need to explore effective and green approaches to enhancing the ability to use chitosan for contaminant removal for practical implementation of this technology. In the present study, glow discharge plasma (GDP), which has thus far been studied
Fenglian Fu et al.
Chemosphere, 66(1), 1-7 (2006-07-11)
Three solid wastes, copper N,N'-bis(dithiocarboxy)piperazine ([CuBDP](n)), copper diethyldithiocarbamate (Cu(DDTC)(2)) and copper dimethyldithiocarbamate (Cu(DMTC)(2)), were prepared and tested as adsorbents to remove Acid Red 73 from wastewater. It was found that the three precipitates all could effectively adsorb the dye but
Chensi Shen et al.
Journal of hazardous materials, 177(1-3), 560-566 (2010-01-09)
A new efficient catalyst, CoTSPc@chitosan, was developed by immobilizing water soluble cobalt (II) tetrasulfophthalocyanine onto adsorbent chitosan microspheres covalently for the heterogeneous catalytic oxidation of C. I. Acid Red 73 with H(2)O(2). The result indicated that the COD removal and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service