208019
Potassium hexacyanoferrate(III)
99%
Synonym(s):
Potassium ferricyanide, Red prussiate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99%
SMILES string
[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N
InChI
1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1
InChI key
MIMJFNVDBPUTPB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2
Supplementary Hazards
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Water research, 47(2), 841-849 (2012-12-04)
Representative and fast monitoring of wastewater influent and effluent biochemical oxygen demand (BOD) is an elusive goal for the wastewater industry and regulatory bodies alike. The present study describes a suitable assay, which incorporates activated sludge as the biocatalyst and
Analytica chimica acta, 724, 92-97 (2012-04-10)
Okadaic acid (OA), a lipophilic phycotoxin is mainly produced by toxigenic dinoflagellates. The need to develop high performing methods for OA analysis able to improve the traditional ones is evident. In this work, a novel experimental methodology for label-free detection
Environmental toxicology and chemistry, 30(5), 1013-1017 (2011-02-11)
The current study deals with the effect of humic substances (HS) on toxicity of solutions of a model inorganic oxidizer, potassium ferricyanide. Chemical reactions responsible for toxicity changes are under consideration. The bioluminescent system of coupled enzymatic reactions catalyzed by
ChemSusChem, 5(6), 1086-1091 (2012-05-10)
To scale-up microbial fuel cells (MFCs), installing multiple unit cells in a common reactor has been proposed; however, there has been a serious potential drop when connecting unit cells in series. To determine the source of the loss, a basic
Journal of pharmaceutical and biomedical analysis, 58, 141-145 (2011-10-15)
In this paper, a novel chemiluminescence (CL) system, 2-phenyl-4, 5-di (2-furyl) imidazole (PDFI)-potassium ferricyanide, for the determination of terbutaline sulfate was described. The method was based on enhancement of CL emission of PDFI-potassium ferricyanide system in the presence of terbutaline
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service