Skip to Content
Merck
All Photos(3)

Key Documents

204412

Sigma-Aldrich

Silver sulfate

99.999% trace metals basis

Synonym(s):

Sulfuric acid disilver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2SO4
CAS Number:
Molecular Weight:
311.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder or crystals

grade

for analytical purposes

Assay

99.999% trace metals basis

form

powder or crystals

reaction suitability

reagent type: catalyst
core: silver

impurities

≤15.0 ppm Trace Metal Analysis

mp

652 °C (lit.)

SMILES string

[Ag+].[Ag+].[O-]S([O-])(=O)=O

InChI

1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

YPNVIBVEFVRZPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service