Skip to Content
Merck
Silver sulfate puriss. p.a., ACS reagent, ≥99.5%

Silver sulfate

Silver sulfate puriss. p.a., ACS reagent, ≥99.5%
Synonym(s):
Silver sulfate, Sulfuric acid disilver(I) salt
Linear Formula:
Ag2SO4
CAS Number:
Molecular Weight:
311.80

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Analyte Suitability
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Technique
Showing 1-6 of 6