Skip to Content
Merck
All Photos(1)

Key Documents

20424

Sigma-Aldrich

(2S,4S)-1-tert-Butoxycarbonyl-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine

≥96.0% (sum of enantiomers, HPLC)

Synonym(s):

(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine, BPPM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H37NO2P2
CAS Number:
Molecular Weight:
553.61
Beilstein:
466510
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥96.0% (sum of enantiomers, HPLC)

form

solid

optical activity

[α]20/D −39±2°, c = 1% in benzene

mp

96-100 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1C[C@H](C[C@H]1CP(c2ccccc2)c3ccccc3)P(c4ccccc4)c5ccccc5

InChI

1S/C34H37NO2P2/c1-34(2,3)37-33(36)35-25-32(39(30-20-12-6-13-21-30)31-22-14-7-15-23-31)24-27(35)26-38(28-16-8-4-9-17-28)29-18-10-5-11-19-29/h4-23,27,32H,24-26H2,1-3H3/t27-,32-/m0/s1

InChI key

BFMKBYZEJOQYIM-UCGGBYDDSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral phosphine ligand for the rhodium catalyzed asymmetric hydrogenation of olefins and ketones. α-Amino acids are obtained in very high optical purity [1]; Hydrogenation of ketones to alcohols[2]; Hydroformylation[3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Parrinello et al.
Journal of the American Chemical Society, 109, 7122-7122 (1987)
I. Ojima et al.
The Journal of Organic Chemistry, 45, 4728-4728 (1980)
Haruka Yonemoto et al.
International journal of molecular sciences, 14(3), 6144-6156 (2013-03-20)
Due to the recent discovery of non-coding RNAs (ncRNAs), multiple sequence alignment (MSA) of those long RNA sequences is becoming increasingly important for classifying and determining the functional motifs in RNAs. However, not only primary (nucleotide) sequences, but also secondary
I. Ojima et al.
Organic Syntheses, 63, 18-18 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service