Skip to Content
Merck
All Photos(3)

Key Documents

200182

Sigma-Aldrich

Nitrazine Yellow

indicator grade, Dye content 85 %

Synonym(s):

2-(2,4-Dinitrophenylazo)-1-hydroxynaphthalene-3,6-disulfonic acid disodium salt, 2-(2,4-Dinitrophenylazo)naphthol-3,6-disulfonic acid disodium salt, Nitrazol Yellow

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N4Na2O11S2
CAS Number:
Molecular Weight:
542.36
Colour Index Number:
14890
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

indicator grade

Assay

>90.0% (HPLC)

form

powder

composition

Dye content, 85%

solubility

H2O: 1 mg/mL

λmax

586 nm

ε (extinction coefficient)

≥35000 at 587-591 nm in 0.1 M NaOH at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1c(\N=N\c2ccc(cc2[N+]([O-])=O)[N+]([O-])=O)c(cc3cc(ccc13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H10N4O11S2.2Na/c21-16-11-3-2-10(32(26,27)28)5-8(11)6-14(33(29,30)31)15(16)18-17-12-4-1-9(19(22)23)7-13(12)20(24)25;;/h1-7,21H,(H,26,27,28)(H,29,30,31);;/q;2*+1/p-2/b18-17+;;

InChI key

XBLIFEQTVVSTIM-QIKYXUGXSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Nitrazine Yellow has been used in dye-protein binding assay.[1]

Biochem/physiol Actions

Nitrazine Yellow ( 2-(2,4-dinitrophenylazo)-1-hydroxynaphthalene-3,6-disulfonic acid disodium salt) is a pH-sensitive azo dye.[2] It changes color from yellow to blue in pH range 6.0–7.0 and is widely used in microscopy.[2][3] Throat swab immersed in nitrazine yellow is used to identify pH variations in the vagina before and after amniotomy.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lien Van der Schueren et al.
Carbohydrate polymers, 91(1), 284-293 (2012-10-10)
Nanofibres functionalised with pH-sensitive dyes could greatly contribute to the development of stimuli-responsive materials. However, the application of biocompatible polymers is vital to allow for their use in (bio)medical applications. Therefore, this paper focuses on the development and characterisation of
Shephali Tagore et al.
Journal of perinatal medicine, 38(6), 609-612 (2010-08-17)
to compare insulin-like growth factor binding protein-1 (IGFBP-1) (non-phosphorylated), placental alpha-microglobulin-1 (PAMG-1) and nitrazine test to diagnose premature rupture of membrane to allow gestation-specific management. we recruited 100 women with signs or symptoms of premature rupture of membranes (PROM), between
[Group hypersensitivity between nitrazine yellow and pellidol (author's transl)].
M Novák
Ceskoslovenska dermatologie, 56(1), 22-24 (1981-02-01)
P Ronnander et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 125, 54-63 (2018-09-18)
A mathematical model was developed to predict the transport of sumatriptan molecules across the skin followed by absorption into the bloodstream. The drug was encapsulated in dissolving polyvinylpyrrolidone-based microneedles shaped in the form of pyramids. Mass balance equations were derived
Why certain dyes are useful for localizing the sentinel lymph node.
Tsopelas C and Sutton R
Journal of Nuclear Medicine, 43, 1377-1382 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service