Skip to Content
Merck
All Photos(1)

Key Documents

196347

Sigma-Aldrich

2-(3-Benzoylphenyl)propionitrile

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H4COC6H5)CN
CAS Number:
Molecular Weight:
235.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

47-53 °C (lit.)

SMILES string

CC(C#N)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H13NO/c1-12(11-17)14-8-5-9-15(10-14)16(18)13-6-3-2-4-7-13/h2-10,12H,1H3

InChI key

RGYOCHMZSLUCNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(3-Benzoylphenyl)propionitrile is the precursor for ketoprofen, widely used anti-inflammatory drug[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purity assay of ketoprofen by high-performance liquid chromatography.
P Pietta et al.
Journal of chromatography, 387, 525-527 (1987-01-30)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service