Skip to Content
Merck
All Photos(1)

Key Documents

195812

Sigma-Aldrich

4-Amino-1-benzylpiperidine

98%

Synonym(s):

1-(Phenylmethyl)-4-piperidinamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$59.10
25 G
$148.00

$59.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$59.10
25 G
$148.00

About This Item

Empirical Formula (Hill Notation):
C12H18N2
CAS Number:
Molecular Weight:
190.28
Beilstein:
146038
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.543 (lit.)

density

0.933 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

NC1CCN(CC1)Cc2ccccc2

InChI

1S/C12H18N2/c13-12-6-8-14(9-7-12)10-11-4-2-1-3-5-11/h1-5,12H,6-10,13H2

InChI key

YUBDLZGUSSWQSS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

4-Amino-1-benzylpiperidine was used in the preparation of:
  • butyl 4-amino-1-piperidineacetate, key intermediate required for the synthesis of butyl 4-[(4-amino-5-chloro-2-methoxybenzoyl)amino]-1-piperidineacetate[1]
  • 5-alkylimino-1,2,4-thiadiazolidine-3-ones, such as 4-ethyl-5-[imino-[1-(phenylmethyl)-4-piperidinyl]]-2-methyl-1,2,4-thiadiazolidin-3-one and 4-benzyl-5-[imino-[1-(phenylmethyl)-4-piperidinyl]]-2-isopropyl-1,2,4-thiadiazolidin-3-one[2]
  • glycyrrhetinic acid derivatives[3]
Reactant for synthesis of:
Highly selective inhibitors of p38a mitogen-activated protein kinase
Antiplasmodial compounds
Dual activity cholinesterase and Aβ-aggregation inhibitors
Muscarinic acetylcholine receptor antagonist and beta 2 adrenoceptor agonists
Photostable near-infrared cyanine dyes
99mTc-labeled piperidine analogues for targeting sigma receptors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federica Prati et al.
ChemMedChem, 11(12), 1284-1295 (2016-02-18)
We discovered a small series of hit compounds that show multitargeting activities against key targets in Alzheimer's disease (AD). The compounds were designed by combining the structural features of the anti-AD drug donepezil with clioquinol, which is able to chelate
J Sakaguchi et al.
Chemical & pharmaceutical bulletin, 49(6), 788-790 (2001-06-20)
A new and facile route for the synthesis of the novel gastrointestinal prokinetic butyl 4-[(4-amino-5-chloro-2-methoxybenzoyl)amino]-1-piperidineacetate (1b), which exhibited potent gastro- and colon-prokinetic activities by oral administration without significant side effects, was established. The key intermediate, butyl 4-amino-1-piperidineacetate (16), was prepared
A Martinez et al.
European journal of medicinal chemistry, 35(10), 913-922 (2000-12-21)
A new family of 1,2,4-thiadiazolidinone derivatives containing the N-benzylpiperidine fragment has been synthesised. The acetylcholinesterase (AChE) inhibitory activity of all compounds was measured using Ellman's method and some of them turned out to be as potent as tacrine. Furthermore, compound
Soo-Jong Um et al.
Bioorganic & medicinal chemistry, 11(24), 5345-5352 (2003-12-04)
To synthesize glycyrrhetinic acid (GA) derivatives (3, 4, 5, 10, 13, 14, 15, and 16), we first removed the ketonic group in the C-11 position, and the carboxylic function at the C-30 position was kept intact, reduced to an alcohol
Josef Dib et al.
Journal of mass spectrometry : JMS, 50(2), 407-417 (2015-03-25)
AdipoR agonists are small, orally active molecules capable of mimicking the protein adiponectin, which represents an adipokine with antidiabetic and antiatherogenic effects. Two adiponectin receptors were reported in the literature referred to as adipoR1 and adipoR2. Activation of these receptors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service