Skip to Content
Merck
All Photos(2)

Key Documents

193372

Sigma-Aldrich

2-(Boc-oxyimino)-2-phenylacetonitrile

99%, for peptide synthesis

Synonym(s):

2-(tert-Butoxycarbonyloxyimino)-2-phenyl­acetonitrile, Boc-ON

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCOON=C(C6H5)CN
CAS Number:
Molecular Weight:
246.26
Beilstein:
2217296
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

2-(Boc-oxyimino)-2-phenylacetonitrile, 99%

Assay

99%

form

powder

mp

85-88 °C (lit.)

application(s)

peptide synthesis

functional group

nitrile
phenyl

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)O\N=C(/C#N)c1ccccc1

InChI

1S/C13H14N2O3/c1-13(2,3)17-12(16)18-15-11(9-14)10-7-5-4-6-8-10/h4-8H,1-3H3/b15-11+

InChI key

QQWYQAQQADNEIC-RVDMUPIBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amino-protecting reagent in peptide synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service