Skip to Content
Merck
All Photos(3)

Key Documents

188735

Sigma-Aldrich

3,5-Dichlorobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$47.10

$47.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$47.10

About This Item

Linear Formula:
Cl2C6H3CH2OH
CAS Number:
Molecular Weight:
177.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$47.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

79-82 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

OCc1cc(Cl)cc(Cl)c1

InChI

1S/C7H6Cl2O/c8-6-1-5(4-10)2-7(9)3-6/h1-3,10H,4H2

InChI key

VSNNLLQKDRCKCB-UHFFFAOYSA-N

Application

3,5-Dichlorobenzyl alcohol was used in the preparation of 3,5-dichlorophenylacetonitrile via reaction with boiling SOCl2, followed by reaction with NaCN[1]. It was also used in the synthesis of tert-butyl 2-cyano-3-(3,5-dichlorophenyl)-propanoate via Iridium-catalysed alkylation of tert-butyl cyanoacetate[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esben R Sørensen et al.
Archiv der Pharmazie, 338(7), 299-304 (2005-07-05)
The HIV-1 inhibitors described in this paper is closely related to 6-(3,5-dimethylbenzyl)-1-(ethoxy methyl)-5-isopropyluracil (GCA-186) an anti-HIV-1 drug that is highly active against both wild type and mutated HIV-1 strains. The two methyl groups on the 6-benzyl moiety have been shown
Iridium catalysed alkylation of tert-butyl cyanoacetate with alcohols under solvent free conditions.
Grigg R, et al.
Tetrahedron, 65(4), 849-854 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service