Skip to Content
Merck
All Photos(2)

Documents

184357

Sigma-Aldrich

(Phenylsulfonyl)acetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2CN
CAS Number:
Molecular Weight:
181.21
Beilstein:
640716
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

112-114 °C (lit.)

SMILES string

O=S(=O)(CC#N)c1ccccc1

InChI

1S/C8H7NO2S/c9-6-7-12(10,11)8-4-2-1-3-5-8/h1-5H,7H2

InChI key

ZFCFFNGBCVAUDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Condensation reaction of (phenylsulfonyl)acetonitrile with benzaldehyde in water in heterogeneous phase in the presence and absence of anionic and cationic surfactants has been studied. Dehydrative alkylation of alcohols with (phenylsulfonyl)acetonitrile under modified Mitsunobu conditions has been investigated.

Application

(Phenylsulfonyl)acetonitrile was used in the synthesis of pyridines, chromenes and thiophene derivatives based on sulfones.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-carbon elongation/annulation of alcohols to nitriles.
Lai J-Y, et al.
Tetrahedron Letters, 36(32), 5691-5694 (1995)
Utility of sulphones in heterocyclic synthesis: Synthesis of some pyridine, chromene and thiophene derivatives.
Fadda AA, et al.
Macromolecules, 5(5), 701-709 (2000)
Condensation reactions in water of active methylene compounds with arylaldehydes. One-pot synthesis of flavonols.
Fringuelli F, et al.
Tetrahedron, 50(39), 11499-11508 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service