Skip to Content
Merck
All Photos(1)

Key Documents

183717

Sigma-Aldrich

2′-Fluoroacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCH3
CAS Number:
Molecular Weight:
138.14
Beilstein:
2041344
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Assay

97%

form

liquid

refractive index

n20/D 1.507 (lit.)

bp

187-189 °C (lit.)

density

1.137 g/mL at 20 °C (lit.)

SMILES string

CC(=O)c1ccccc1F

InChI

1S/C8H7FO/c1-6(10)7-4-2-3-5-8(7)9/h2-5H,1H3

InChI key

QMATYTFXDIWACW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The enantioselective reduction of 2′-fluoroacetophenone has been investigated[1].

Application

2′-Fluoroacetophenone was used as starting reagent in the synthesis of ascididemin[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ida Nymann Petersen et al.
Chemical communications (Cambridge, England), 48(72), 9092-9094 (2012-08-07)
A new and convergent synthesis of ascididemin is presented. Using an anionic cascade ring closure as the key step, this natural product is obtained in 45% overall yield in just 6 steps starting from 2'-fluoroacetophenone. This new approach was extended
The enantioselective reduction of 2'-fluoroacetophenone utilizing a simplified CBS-reduction procedure.
Garrett CE, et al.
Tetrahedron Asymmetry, 13(13), 1347-1349 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service